missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Isopropylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64695.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469076

  • £74.73 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Isopropylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
338.32
MFCD26960709

Specifications

1-(3-Isopropylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
338.32
1g
97%
MFCD26960709
Videos
SDS