missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Iodobenzyl)piperidine, ≥97%, Maybridge™

£60.28 - £519.00

Chemical Identifiers

CAS 859850-87-6
Molecular Formula C12H16IN
Molecular Weight (g/mol) 301.171
MDL Number MFCD06797854
InChI Key DCRZUICHKJGHQK-UHFFFAOYSA-N
Synonym 1-3-iodobenzyl piperidine, 1-3-iodophenyl methyl piperidine, piperidine,1-3-iodophenyl methyl, 1-3-iodo-benzyl-piperidine
PubChem CID 17750314
IUPAC Name 1-[(3-iodophenyl)methyl]piperidine
SMILES C1CCN(CC1)CC2=CC(=CC=C2)I
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10041434
SDS View Documents
Maybridge
CC45211CB
250mg
£60.28
250mg
View alternative products
Save Up to 
»
Estimated Shipment
16-06-2021
Please to purchase this item. Need a web account? Register with us today!
10670084
SDS View Documents
Maybridge
CC45211DA
1g
£155.00
1g
View alternative products
Save Up to 
»
Estimated Shipment
16-06-2021
Please to purchase this item. Need a web account? Register with us today!
10210275
SDS View Documents
Maybridge
CC45211DE
5g
£519.00
5g
View alternative products
Save Up to 
»
Estimated Shipment
16-06-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

859850-87-6
301.171
DCRZUICHKJGHQK-UHFFFAOYSA-N
17750314
C1CCN(CC1)CC2=CC(=CC=C2)I
C12H16IN
MFCD06797854
1-3-iodobenzyl piperidine, 1-3-iodophenyl methyl piperidine, piperidine,1-3-iodophenyl methyl, 1-3-iodo-benzyl-piperidine
1-[(3-iodophenyl)methyl]piperidine

Specifications

1-(3-Iodobenzyl)piperidine
250mg
96.5
97%
MFCD06797854
DCRZUICHKJGHQK-UHFFFAOYSA-N
1-[(3-iodophenyl)methyl]piperidine
17750314
97%
Amber glass bottle
859850-87-6
100.0
C12H16IN
1-3-iodobenzyl piperidine, 1-3-iodophenyl methyl piperidine, piperidine,1-3-iodophenyl methyl, 1-3-iodo-benzyl-piperidine
C1CCN(CC1)CC2=CC(=CC=C2)I
301.171
301.17
SDS