missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61473.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466536

  • £68.26 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
298.24
MFCD03760974

Specifications

1-(3-Fluorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
1g
97%
MFCD03760974
White
Solid
298.24
Videos
SDS