missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64120.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15428716

  • £66.46 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
314.24
MFCD05077945

Specifications

1-(3-Fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
314.24
1g
97%
MFCD05077945
Videos
SDS