Alfa Aesar™ 1-(3-Fluorophenyl)-3-(4-fluorophenyl)urea, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H62961.MD

UNSPSC: 12352116

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15488046

Quantity Price
1 £ 14.1 / 250mg
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-(3-Fluorophenyl)-3-(4-fluorophenyl)urea
Formula Weight 248.23
MDL Number MFCD07080167

Specification

Formula Weight 248.23
Physical Form Solid
MDL Number MFCD07080167
Chemical Name or Material 1-(3-Fluorophenyl)-3-(4-fluorophenyl)urea
Quantity 250mg
Assay 97%