missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Fluorobenzyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64002.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498626

  • £70.28 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Fluorobenzyl)-3-[4-(trifluoromethoxy)phenyl]urea
328.26
MFCD27665241

Specifications

1-(3-Fluorobenzyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
328.26
1g
97%
MFCD27665241
Videos
SDS