missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Ethylphenyl)ethan-1-one, 95%, Maybridge

Brand:  Maybridge TL00574DA

Code : 31

Additional Details : CAS Number : 22699-70-3 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10308622

  • £37.00 / 1g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Ethylphenyl)ethan-1-one
95%
C2H5C6H4COCH3
95%
C10H12O
148.2

Specifications

1-(3-Ethylphenyl)ethan-1-one
22699-70-3
Amber Glass Bottle
95%
C2H5C6H4COCH3
1g
0.9300g/mL
95%
C10H12O
148.2
Videos
SDS