missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Ethylphenyl)ethan-1-one, 95%, Maybridge

Brand:  Maybridge TL00574EA

Code : 31

Additional Details : CAS Number : 22699-70-3 Weight : 0.06000kg

Quantity 10g

 View more versions of this product

Product Code. 10231423

  • £213.00 / 10g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Ethylphenyl)ethan-1-one
95%
C2H5C6H4COCH3
95%
C10H12O
148.2

Specifications

1-(3-Ethylphenyl)ethan-1-one
22699-70-3
95%
C2H5C6H4COCH3
10g
95%
C10H12O
148.2
Videos
SDS