missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dithiane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1,3-Dithiane is used in umpolung reactions such as the Corey-Seebach reaction. It acts as a reagent for the deoxygenation of sulfoxides to their corresponding sulfides. Further, it is used as a protecting group for carbonyl compounds used in organic synthesis. It serves as a useful labeled synthon. In addition to this, it is used in the preparation of 2-ethyl-[1,3]dithiane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Dithiane is used in umpolung reactions such as the Corey-Seebach reaction. It acts as a reagent for the deoxygenation of sulfoxides to their corresponding sulfides. Further, it is used as a protecting group for carbonyl compounds used in organic synthesis. It serves as a useful labeled synthon. In addition to this, it is used in the preparation of 2-ethyl-[1,3]dithiane.
Solubility
Soluble in benzene, ether, chloroform and terahydrofuran. Slightly soluble in water.
Notes
Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
1,3-Dithiane is used in umpolung reactions such as the Corey-Seebach reaction. It acts as a reagent for the deoxygenation of sulfoxides to their corresponding sulfides. Further, it is used as a protecting group for carbonyl compounds used in organic synthesis. It serves as a useful labeled synthon. In addition to this, it is used in the preparation of 2-ethyl-[1,3]dithiane.
Solubility
Soluble in benzene, ether, chloroform and terahydrofuran. Slightly soluble in water.
Notes
Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
Specifications
Melting Point | 52°C to 55°C |
Boiling Point | 207°C to 208°C |
Flash Point | 90°C (194°F) |
Odor | Stench |
Quantity | 5 g |
UN Number | UN3335 |
Beilstein | 102534 |
Solubility Information | Soluble in benzene,ether,chloroform and terahydrofuran. Slightly soluble in water. |
Formula Weight | 120.24 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'