missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dithiane, 97%, Thermo Scientific Chemicals

Catalog Number p-3778466
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
505-23-7
C4H8S2
120.23
MFCD00006654
WQADWIOXOXRPLN-UHFFFAOYSA-N
1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk
10451
CHEBI:47845
1,3-dithiane
C1CSCSC1
This item is not returnable. View return policy
505-23-7
C4H8S2
120.23
MFCD00006654
WQADWIOXOXRPLN-UHFFFAOYSA-N
1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk
10451
CHEBI:47845
1,3-dithiane
C1CSCSC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 49°C to 54°C
Color White to Beige
Density 90g/mL
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Fieser 02,182; 03,135; 04,216; 05,287; 06,248; 07,142; 14,157; 16,161; 01,208; 02,108; 12,158; 15,112
Formula Weight 120.23
Percent Purity 97%
Physical Form Crystals, Needles or Powder
Chemical Name or Material 1, 3-Dithiane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'