Learn More
1,3-Dipropyl-8-phenylxanthine, Tocris Bioscience™
A1 selective antagonist
Brand:  Tocris (Bio-Techne) 048610
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487723
  • £128.25 / 10mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Selective A1 adenosine antagonist.
Specifications
Chemical Identifiers
85872-53-3
312.373
128784
C17H20N4O2
CLIGSMOZKDCDRZ-UHFFFAOYSA-N
Specifications
1, 3-Dipropyl-8-phenylxanthine
85872-53-3
C17H20N4O2
CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=CC=C3
312.373
312.37
10mg
>98%
CLIGSMOZKDCDRZ-UHFFFAOYSA-N
8-phenyl-1,3-dipropyl-7H-purine-2,6-dione
128784
Videos