missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Diphenylguanidine, primary standard, 99+%

Primary material for standardizing acids

Brand:  Alfa Aesar™ 031395.09

Code : NEW

Additional Details : CAS Number : 102-06-7 Weight : 0.01000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11317468

  • £11.40 / 10g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

1,3-Diphenylguanidine
Primary standard
MFCD00001758
1875653
N,N'-Diphenylguanidine; Melaniline
102-06-7
211.27
UN3077
14,3324

Specifications

1,3-Diphenylguanidine
102-06-7
Characteristic
≥99%
MFCD00001758
1875653
N,N'-Diphenylguanidine; Melaniline
170°C (dec.)
10g
Primary standard
1.130g/mL
Powder
211.27
UN3077
14,3324
148° to 150°C
Pale Cream
Videos
SDS