missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Primary material for standardizing acids. Diphenylguanidine is used as a medium of natural and synthetic rubber accelerator. It acts as a complexing agent for the detection of metals and organic bases. Further, it is used as a secondary foam stabilizer in the silicoflouride foam process. It is actively involved in the manufacture of rubber sheet, rubber shoes and other rubber industrial products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 148°C to 150°C |
Color | Cream |
Density | 1.130 g/mL |
Boiling Point | 170°C (decomposition) |
Odor | Characteristic |
Quantity | 50 g |
UN Number | UN3077 |
Beilstein | 1875653 |
Merck Index | 14,3324 |
Solubility Information | Soluble in water,ethanol,carbon tetrachloride,chloroform,toluene and ethyl ether. Slightly soluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'