missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylacetone, 99%, ACROS Organics™

£35.60 - £427.20

Chemical Identifiers

CAS 102-04-5
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.27
InChI Key YFKBXYGUSOXJGS-UHFFFAOYSA-N
Synonym 1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
PubChem CID 7593
IUPAC Name 1,3-diphenylpropan-2-one
SMILES C1=CC=C(C=C1)CC(=O)CC2=CC=CC=C2
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10011040 SDS View Documents Acros Organics
117170250
25g Glass bottle £35.60
25g View alternative products
Save Up to 
»
EU Stock 8 Please to purchase this item. Need a web account? Register with us today!
10051070 SDS View Documents Acros Organics
117171000
100g Glass bottle £94.40
100g View alternative products
Save Up to 
»
EU Stock 40 Please to purchase this item. Need a web account? Register with us today!
11994841 SDS View Documents Acros Organics
117175000
500g Glass bottle £427.20
500g View alternative products
Save Up to 
»
EU Stock 7 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

102-04-5
210.27
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
1,3-diphenylpropan-2-one
C15H14O
YFKBXYGUSOXJGS-UHFFFAOYSA-N
7593
C1=CC=C(C=C1)CC(=O)CC2=CC=CC=C2

Specifications

1, 3-Diphenylacetone
>110°C
Glass bottle
330.0°C
30.0°C to 34.0°C
102-04-5
100.0
C15H14O
07,445
1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
YFKBXYGUSOXJGS-UHFFFAOYSA-N
1,3-diphenylpropan-2-one
7593
Liquid or Crystalline Mass
1.0400g/mL
Authentic
1.04
Yellow
25g
98.5
98.5% min. (GC)
C6H5CH2COCH2C6H5
01,198; 04,123
Solubility in water: insoluble. Other solubilities: soluble ethanol, diethyl ether, petroleum ether
C1=CC=C(C=C1)CC(=O)CC2=CC=CC=C2
210.27
210.27
99%