missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Catalog Number p-2694774
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
5356-71-8
C15H13N3
235.29
MFCD00084878
SXOFMEWDEKEVJU-UHFFFAOYSA-N
1,3-diphenyl-1h-pyrazol-5-amine, 5-amino-1,3-diphenylpyrazole, 1h-pyrazol-5-amine, 1,3-diphenyl, pyrazole, 5-amino-1,3-diphenyl, 5-amino-1,3-diphenyl-1h-pyrazole, 1,3-diphenyl-1h-pyrazol-5-ylamine, 1,3-diphenylpyrazole-5-ylamine, maybridge1_004139, acmc-1apw6, cbmicro_003752
199969
2,5-diphenylpyrazol-3-amine
C1=CC=C(C=C1)C2=NN(C(=C2)N)C3=CC=CC=C3
This item is not returnable. View return policy
5356-71-8
C15H13N3
235.29
MFCD00084878
SXOFMEWDEKEVJU-UHFFFAOYSA-N
1,3-diphenyl-1h-pyrazol-5-amine, 5-amino-1,3-diphenylpyrazole, 1h-pyrazol-5-amine, 1,3-diphenyl, pyrazole, 5-amino-1,3-diphenyl, 5-amino-1,3-diphenyl-1h-pyrazole, 1,3-diphenyl-1h-pyrazol-5-ylamine, 1,3-diphenylpyrazole-5-ylamine, maybridge1_004139, acmc-1apw6, cbmicro_003752
199969
2,5-diphenylpyrazol-3-amine
C1=CC=C(C=C1)C2=NN(C(=C2)N)C3=CC=CC=C3
CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 235.29
Percent Purity 97%
Chemical Name or Material 1, 3-diphenyl-1H-pyrazol-5-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'