missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 235.29 |
Percent Purity | 97% |
Chemical Name or Material | 1, 3-diphenyl-1H-pyrazol-5-amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'