missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenoxybenzene, 99%, ACROS Organics™

Brand:  Acros Organics 408840250

Code : 29

Additional Details : CAS Number : 3379-38-2 Weight : 0.07500kg

Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10548322

  • £103.10 / 25g


Estimated Shipment
19-07-2018
Add to basket

Specifications

Chemical Identifiers

1, 3-Diphenoxybenzene
99%
262.3
99%
C18H14O2

Specifications

1, 3-Diphenoxybenzene
Authentic
Crystals or Crystalline Powder
99%
262.3
White
3379-38-2
Glass bottle
99%
C18H14O2
58.0° to 61.0°C
25g
Videos
SDS