missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%, Thermo Scientific Chemicals

Catalog Number p-7081130
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
22572-40-3
C9H20N3+
170.28
MFCD00011833
JHMDMCSGQQLHNA-UHFFFAOYSA-N
3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
122243
3-(ethyliminomethylideneamino)propyl-trimethylazanium
CCN=C=NCCC[N+](C)(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
22572-40-3
C9H20N3+
170.28
MFCD00011833
JHMDMCSGQQLHNA-UHFFFAOYSA-N
3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
122243
3-(ethyliminomethylideneamino)propyl-trimethylazanium
CCN=C=NCCC[N+](C)(C)C

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

Solubility
Soluble in water.

Notes
Keep in a dry, cool and well-ventilated place. Keep container tightly closed. Incompatible with acids, strong oxidizing agents, moist air and water.
TRUSTED_SUSTAINABILITY
Melting Point 90°C to 92°C
Quantity 1 g
Beilstein 4162760
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 297.17
Percent Purity 98%
Chemical Name or Material 1- (3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'