missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals

Catalog Number p-117959
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
80-73-9
C5H10N2O
114.15
MFCD00003188
CYSGHNMQYZDMIA-UHFFFAOYSA-N
1,3-dimethyl-2-imidazolidinone, n,n'-dimethylethyleneurea, dimethyl imidazolidinone, n,n'-dimethylimidazolidinone, 1,3-dimethylimidazolidone, rhonite 1, dmeu, karbomos tsem, 2-imidazolidinone, 1,3-dimethyl, 1,3-dimethylethyleneurea
6661
1,3-dimethylimidazolidin-2-one
CN1CCN(C)C1=O
This item is not returnable. View return policy
80-73-9
C5H10N2O
114.15
MFCD00003188
CYSGHNMQYZDMIA-UHFFFAOYSA-N
1,3-dimethyl-2-imidazolidinone, n,n'-dimethylethyleneurea, dimethyl imidazolidinone, n,n'-dimethylimidazolidinone, 1,3-dimethylimidazolidone, rhonite 1, dmeu, karbomos tsem, 2-imidazolidinone, 1,3-dimethyl, 1,3-dimethylethyleneurea
6661
1,3-dimethylimidazolidin-2-one
CN1CCN(C)C1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 8.2°C
Density 1.0500g/mL
Boiling Point 225.5°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4710 to 1.4730
Quantity 100 g
Fieser 11,202; 15,145
Merck Index 15, 3273
Specific Gravity 1.05
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol, ethanol, 2-propanol, ether,, methylene chloride, dmf, acetone, ketone and, chloroform
Viscosity 1.944 mPa.s (25°C)
Formula Weight 114.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3-Dimethyl-2-imidazolidinone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Dimethyl-2-imidazolidinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'