missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethyl-1H-pyrazole-5-carbaldehyde, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 1,3-dimethyl-1h-pyrazole-5-carbaldehyde, 1,3-dimethylpyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1,3-dimethyl, 1,3-dimethyl-5-formyl-1h-pyrazole, 2,5-dimethyl-2h-pyrazole-3-carbaldehyde, 1,3-dimethylpyrazole-5-carboxaldehyde, 1,3-dimethyl-1h-pyrazole-5-carboxaldehyde, 2,5-dimethyl-3-pyrazolecarboxaldehyde, 1,3-dimethyl-1h-pyrazol-5-carbaldehyde, 1,3-dimethyl-1 h-pyrazole-5-carbaldehyde |
Formula Weight | 124.14 |
Percent Purity | 90% |
Chemical Name or Material | 1, 3-Dimethyl-1H-pyrazole-5-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'