missing translation for 'onlineSavingsMsg'
Learn More

(1,3-Dimethyl-1H-pyrazol-5-yl)methylamine, 97%, Maybridge

Brand:  Maybridge CC12513CB

Code : 31

Additional Details : CAS Number : 499770-63-7 Weight : 0.05025kg Transport : UN number : 3259 Chem class : 1b Pack group : III

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10130632

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

(1, 3-Dimethyl-1H-pyrazol-5-yl)methylamine
95%
125.17
95%
C6H11N3

Specifications

(1, 3-Dimethyl-1H-pyrazol-5-yl)methylamine
499770-63-7
95%
C6H11N3
250mg
Amber Glass Bottle
95%
125.17
Videos
SDS