missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 10 g |
Formula Weight | 111.15 |
Percent Purity | 97% |
Chemical Name or Material | 1, 3-Dimethyl-1H-pyrazol-5-amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'