missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethoxybenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-114095
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
151-10-0
C8H10O2
138.17
MFCD00008384
DPZNOMCNRMUKPS-UHFFFAOYSA-N
m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
9025
1,3-dimethoxybenzene
COC1=CC(OC)=CC=C1
This item is not returnable. View return policy
151-10-0
C8H10O2
138.17
MFCD00008384
DPZNOMCNRMUKPS-UHFFFAOYSA-N
m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu
9025
1,3-dimethoxybenzene
COC1=CC(OC)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Brown
Density 1.0500g/mL
Boiling Point 85.0°C to 87.0°C (7.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(OCH3)2
Refractive Index 1.5230 to 1.5250
Quantity 100 g
Beilstein 06, 813
Specific Gravity 1.05
Solubility Information Solubility in water: immiscible
Formula Weight 138.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 3-Dimethoxybenzene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'