missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one, 97%, Thermo Scientific Chemicals

Catalog Number p-4509918
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2147-83-3
C12H14N3O
216.26
MFCD00044827
YFEOSTXFQCDCAR-UHFFFAOYSA-O
1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazole-2-one, 1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazol-2-one, 3-1,2,3,6-tetrahydropyridin-4-yl-1h-benzimidazol-2-one, 1-1,2,3,6-tetrahydro-4-pyridyl-2-benzimidazolinone, 1-1,2,3,6-tetrahydropyridin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-4-1,2,5,6-tetrahydropyridyl-3-hydrobenzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-3h-1,3-benzodiazol-2-one, benzoimidazol-2-one, 1-1,2,3,6-tetrahydro-pyridin-4-yl-1,3-dihydro
75070
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-benzimidazol-2-one
O=C1NC2=CC=CC=C2N1C1=CC[NH2+]CC1
This item is not returnable. View return policy
2147-83-3
C12H14N3O
216.26
MFCD00044827
YFEOSTXFQCDCAR-UHFFFAOYSA-O
1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazole-2-one, 1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazol-2-one, 3-1,2,3,6-tetrahydropyridin-4-yl-1h-benzimidazol-2-one, 1-1,2,3,6-tetrahydro-4-pyridyl-2-benzimidazolinone, 1-1,2,3,6-tetrahydropyridin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-4-1,2,5,6-tetrahydropyridyl-3-hydrobenzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-3h-1,3-benzodiazol-2-one, benzoimidazol-2-one, 1-1,2,3,6-tetrahydro-pyridin-4-yl-1,3-dihydro
75070
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-benzimidazol-2-one
O=C1NC2=CC=CC=C2N1C1=CC[NH2+]CC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 195.0°C to 200.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,dichloromethane and acetone
Formula Weight 215.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'