missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoro-2-propanol, 98%, ACROS Organics™

Brand:  Acros Organics 175560010

Code : 29

Additional Details : CAS Number : 453-13-4 Weight : 0.05100kg Transport : UN number : 1986 Chem class : 3 Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10322913

  • £25.20 / 1g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

1, 3-Difluoro-2-propanol
453-13-4
100.0
98%
FCH2CH(OH)CH2F
MFCD00000452
99%
98.5
≥97.5% (GC)
C3H6F2O
96.08

Specifications

1, 3-Difluoro-2-propanol
Clear yellow to brownish
Authentic
453-13-4
100.0
1.3720 to 1.3750 (20°C, 589nm)
≥97.5% (GC)
C3H6F2O
96.08
1.2400g/mL
1g
99%
98.5
Glass bottle
Liquid
98%
FCH2CH(OH)CH2F
MFCD00000452
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,3-Difluoro-2-propanol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS