missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Difluoro-2-propanol, 98%

£15.40 - £46.40

Chemical Identifiers

CAS 453-13-4
Molecular Formula C3H6F2O
Molecular Weight (g/mol) 96.077
MDL Number MFCD00000452
InChI Key PVDLUGWWIOGCNH-UHFFFAOYSA-N
Synonym 1,3-difluoro-2-propanol, 2-propanol, 1,3-difluoro, 1,3-difluoroisopropanol, 2-propanol,3-difluoro, wln: f1yq1f, sgqdlibrbrwup@, acmc-20ap4b, 1,3-difluoropropane-2-ol, 1,3-difluoro-propan-2-ol, 1,3-difluoro-2-hydroxypropane
PubChem CID 67985
IUPAC Name 1,3-difluoropropan-2-ol
SMILES C(C(CF)O)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11321367 SDS View Documents Alfa Aesar™
L01646.03
1g £15.40
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 28-10-2019
Please to purchase this item. Need a web account? Register with us today!
11331367 SDS View Documents Alfa Aesar™
L01646.06
5g £46.40
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-11-2019
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

453-13-4
96.077
PVDLUGWWIOGCNH-UHFFFAOYSA-N
67985
C(C(CF)O)F
C3H6F2O
MFCD00000452
1,3-difluoro-2-propanol, 2-propanol, 1,3-difluoro, 1,3-difluoroisopropanol, 2-propanol,3-difluoro, wln: f1yq1f, sgqdlibrbrwup@, acmc-20ap4b, 1,3-difluoropropane-2-ol, 1,3-difluoro-propan-2-ol, 1,3-difluoro-2-hydroxypropane
1,3-difluoropropan-2-ol

Specifications

1g
42°C (107°F)
453-13-4
MFCD00000452
1732050
PVDLUGWWIOGCNH-UHFFFAOYSA-N
1,3-difluoropropan-2-ol
67985
98%
1,3-Difluoro-2-propanol
127°C
C3H6F2O
UN1986
1,3-difluoro-2-propanol, 2-propanol, 1,3-difluoro, 1,3-difluoroisopropanol, 2-propanol,3-difluoro, wln: f1yq1f, sgqdlibrbrwup@, acmc-20ap4b, 1,3-difluoropropane-2-ol, 1,3-difluoro-propan-2-ol, 1,3-difluoro-2-hydroxypropane
C(C(CF)O)F
96.077
96.08