missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 96%, ACROS Organics™

Brand:  Acros Organics 215280250

Code : 29

Additional Details : CAS Number : 69304-37-6 Weight : 0.07500kg Transport : UN number : 2987 Chem class : 1a Pack group : II


Quantity 25mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10513324

  • £141.40 / 25mL


Estimated Shipment
20-02-2018
Add to basket


Specifications

Chemical Identifiers

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
69304-37-6
100.0
96%
[(CH3)2CH]2Si(Cl)OSi(Cl)[CH(CH3)2]2
MFCD00009655
96%
95.0
96%
C12H28Cl2OSi2
315.42
TIPSCl

Specifications

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
76°C
Clear colorless
96%
Authentic
100.0
1.4533 to 1.4553
96%
C12H28Cl2OSi2
315.42
16,125; 17,121
1.0000g/mL
120.0°C (15.0 mmHg)
25mL
69304-37-6
95.0
Glass bottle
Liquid
96%
[(CH3)2CH]2Si(Cl)OSi(Cl)[CH(CH3)2]2
MFCD00009655
TIPSCl
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS