missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromopropane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | °C to 34°C |
Density | 1.99 |
Boiling Point | 167°C |
Flash Point | 54°C (130°F) |
Assay Percent Range | 98% |
Linear Formula | Br(CH2)3Br |
Refractive Index | 1.523 |
Quantity | 250 g |
UN Number | UN1993 |
Beilstein | 635662 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'