missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromoacetone, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications
Melting Point | 29°C to 30°C |
Color | White to Yellow |
Boiling Point | 97°C to 98°C (21 mmHg) |
Flash Point | 102°C (216°F) |
Quantity | 25 g |
UN Number | UN1759 |
Beilstein | 1740389 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in acetone,chloroform,dichloromethane and methanol. |
Formula Weight | 215.87 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'