missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Dibromo-5-(trifluoromethoxy)benzene, 97%

Brand:  Alfa Aesar™ H64115.14

Code : NEW

Additional Details : CAS Number : 207226-31-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15451169

  • £41.80 / 25g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

1,3-Dibromo-5-(trifluoromethoxy)benzene
207226-31-1
319.9
MFCD00075280

Specifications

1,3-Dibromo-5-(trifluoromethoxy)benzene
1.5080
97%
MFCD00075280
83° to 85°C (20mmHg) (lit.)
207226-31-1
Liquid
319.9
Colorless
25g
Videos
SDS