missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-2-propanol, 95%, ACROS Organics™

£50.90 - £148.90

Chemical Identifiers

CAS 96-21-9
Molecular Formula C3H6Br2O
Molecular Weight (g/mol) 217.89
MDL Number MFCD00000216
InChI Key KIHQZLPHVZKELA-UHFFFAOYSA-N
Synonym 1,3-dibromo-2-propanol, 2-propanol, 1,3-dibromo, 1,3-dibromohydrin, glycerol 1,3-dibromohydrin, 2-hydroxy-1,3-dibromopropane, 1,3-dibromo-2-hydroxypropane, alpha-dibromohydrin, .alpha.-dibromohydrin, glycerol alpha,gamma-dibromohydrin, glycerol alpha,gamma-dibromohydrine
PubChem CID 7287
IUPAC Name 1,3-dibromopropan-2-ol
SMILES C(C(CBr)O)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10055882 SDS View Documents Acros Organics
406540250
25g Glass bottle £50.90
25g View alternative products
Save Up to 
»
EU Stock 4 Please to purchase this item. Need a web account? Register with us today!
10569082 SDS View Documents Acros Organics
406541000
100g Glass bottle £148.90
100g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 19-12-2019
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

96-21-9
217.89
KIHQZLPHVZKELA-UHFFFAOYSA-N
7287
C(C(CBr)O)Br
C3H6Br2O
MFCD00000216
1,3-dibromo-2-propanol, 2-propanol, 1,3-dibromo, 1,3-dibromohydrin, glycerol 1,3-dibromohydrin, 2-hydroxy-1,3-dibromopropane, 1,3-dibromo-2-hydroxypropane, alpha-dibromohydrin, .alpha.-dibromohydrin, glycerol alpha,gamma-dibromohydrin, glycerol alpha,gamma-dibromohydrine
1,3-dibromopropan-2-ol

Specifications

1, 3-Dibromo-2-propanol
46°C
Glass bottle
2.13
Brown or Colorless to Yellow
pract., 85%
94.0
95%
CH2BrCHOHCH2Br
01, 365
Solubility in water: soluble. Other solubilities: very soluble in acetone,ethyl ether and ethanol
C(C(CBr)O)Br
217.89
217.89
≥94%
2.1300g/mL
Authentic
1.5500 to 1.554
219°C
25g
96-21-9
100.0
C3H6Br2O
MFCD00000216
1,3-dibromo-2-propanol, 2-propanol, 1,3-dibromo, 1,3-dibromohydrin, glycerol 1,3-dibromohydrin, 2-hydroxy-1,3-dibromopropane, 1,3-dibromo-2-hydroxypropane, alpha-dibromohydrin, .alpha.-dibromohydrin, glycerol alpha,gamma-dibromohydrin, glycerol alpha,gamma-dibromohydrine
KIHQZLPHVZKELA-UHFFFAOYSA-N
1,3-dibromopropan-2-ol
7287
Liquid
Safety and Handling

Safety and Handling

Product Identifier
  • 1,3-Dibromo-2-propanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.