missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Diazaspiro[4.4]nonane-2,4-dione, 95%

Brand:  Alfa Aesar™ H64444.06

Code : NEW

Additional Details : CAS Number : 699-51-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15451659

  • £152.64 / 5g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

1,3-Diazaspiro[4.4]nonane-2,4-dione
154.17
135190
699-51-4
MFCD00022403
5,5-Tetramethylenehydantoin

Specifications

1,3-Diazaspiro[4.4]nonane-2,4-dione
199° to 205°C
95%
154.17
135190
699-51-4
Solid
5g
MFCD00022403
5,5-Tetramethylenehydantoin
Videos
SDS