missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane, 98%, Thermo Scientific Chemicals

Catalog Number p-7081787
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
109-76-2
C3H10N2
74.127
MFCD00008228
XFNJVJPLKCPIBV-UHFFFAOYSA-N
1,3-diaminopropane, 1,3-propanediamine, trimethylenediamine, 1,3-propylenediamine, propandiamine, alpha,omega-propanediamine, unii-cb3isl56kg, ccris 4054, 1,3-propane diamine, cb3isl56kg
428
CHEBI:15725
propane-1,3-diamine
C(CN)CN
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
109-76-2
C3H10N2
74.127
MFCD00008228
XFNJVJPLKCPIBV-UHFFFAOYSA-N
1,3-diaminopropane, 1,3-propanediamine, trimethylenediamine, 1,3-propylenediamine, propandiamine, alpha,omega-propanediamine, unii-cb3isl56kg, ccris 4054, 1,3-propane diamine, cb3isl56kg
428
CHEBI:15725
propane-1,3-diamine
C(CN)CN

1,3-Diaminopropane is commonly used in the preparation of cholinesterase inhibitors and thrombosis inhibitors. It is also used as a polymer cross linkage agent, polyurethane extender and spray coatings. It is involved in the preparation of agrochemicals and pharmaceutical intermediates. Further, it acts as a building block in the synthesis of heterocyclic compounds, which finds application in textile finishing. In addition to this, it is used as a ligand and form coordination complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diaminopropane is commonly used in the preparation of cholinesterase inhibitors and thrombosis inhibitors. It is also used as a polymer cross linkage agent, polyurethane extender and spray coatings. It is involved in the preparation of agrochemicals and pharmaceutical intermediates. Further, it acts as a building block in the synthesis of heterocyclic compounds, which finds application in textile finishing. In addition to this, it is used as a ligand and form coordination complexes.

Solubility
Miscible with water.

Notes
Hygroscopic. Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY
Melting Point -12°C
Density 0.888
pH >12
Boiling Point 134°C to 136°C
Flash Point 48°C (118°F)
Odor Amine-like
Refractive Index 1.4573
Quantity 100 mL
UN Number UN2734
Beilstein 605277
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 74.13
Percent Purity 98%
Chemical Name or Material 1,3-Diaminopropane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'