missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butylbenzene, 99%, Thermo Scientific Chemicals

Catalog Number 13448638
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1014-60-4
C14H22
190.33
MFCD00008830
ILNDSSCEZZFNGE-UHFFFAOYSA-N
1,3-di-tert-butylbenzene, benzene, 1,3-bis 1,1-dimethylethyl, benzene, m-di-tert-butyl, m-di-tert-butylbenzene, 1,3-di-t-butylbenzene, 1,3-ditert-butyl-benzene, 1,3-ditertiarybutylbenzene, 1,3-di-tert-butyl-benzene, acmc-1bo58, 1,3-bis tert-butyl benzene
136810
1,3-ditert-butylbenzene
CC(C)(C)C1=CC(=CC=C1)C(C)(C)C
This item is not returnable. View return policy
1014-60-4
C14H22
190.33
MFCD00008830
ILNDSSCEZZFNGE-UHFFFAOYSA-N
1,3-di-tert-butylbenzene, benzene, 1,3-bis 1,1-dimethylethyl, benzene, m-di-tert-butyl, m-di-tert-butylbenzene, 1,3-di-t-butylbenzene, 1,3-ditert-butyl-benzene, 1,3-ditertiarybutylbenzene, 1,3-di-tert-butyl-benzene, acmc-1bo58, 1,3-bis tert-butyl benzene
136810
1,3-ditert-butylbenzene
CC(C)(C)C1=CC(=CC=C1)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 10.0°C to 11.0°C
Density 0.8600g/mL
Boiling Point 106.0°C to 107.0°C (18.0 mmHg)
Flash Point 84°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4C(CH3)3
Quantity 25 g
Specific Gravity 0.86
Formula Weight 190.33
Percent Purity 99%
Chemical Name or Material 1,3-Di-tert-butylbenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'