Alfa Aesar™ 1,3-Di-tert-butyl-2-methoxybenzene, 99%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H37487.18

UNSPSC: 12352100

Code: NEW

Additional Details:
Additional Details: CAS Number: 1516-95-6 Weight: 0.05000kg


Quantity 50g

Product Code. 15427787

Quantity Price
1 £ 76.43 / 50g
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1,3-Di-tert-butyl-2-methoxybenzene
CAS 1516-95-6
Formula Weight 220.35
MDL Number MFCD20483412
Synonym 2,6-Di-tert-butyl anisole

Specification

Formula Weight 220.35
Synonym 2,6-Di-tert-butyl anisole
Chemical Name or Material 1,3-Di-tert-butyl-2-methoxybenzene
Boiling Point 124°C (9mmHg)
Color Colorless to pale yellow
MDL Number MFCD20483412
Quantity 50g
Assay 99%
CAS 1516-95-6
Physical Form Liquid
Documentation

Technical documents