missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane, 94%, Thermo Scientific™

Catalog Number p-7030491
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
18055-70-4
C18H26OSi2
314.58
MFCD09266163
MEQYEVSZYLVDNX-UHFFFAOYSA-N
sym-tetramethyl-di-p-tolyl disiloxane, 1,3-di p-tolyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis p-tolyl-1,1,3,3-tetramethyl disiloxane, disiloxane, 1,1,3,3-tetramethyl-1,3-bis 4-methylphenyl, dimethyl 4-methylphenyl silyl oxy dimethyl 4-methylphenyl silane, amtsi070, 1,3-di-p-tolyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis 4-methylphenyl-1,1,3,3-tetramethyldisiloxane, 1,3-di 4-methylphenyl-1,1,3,3-tetramethyldisiloxane
519510
{[dimethyl(4-methylphenyl)silyl]oxy}dimethyl(4-methylphenyl)silane
CC1=CC=C(C=C1)[Si](C)(C)O[Si](C)(C)C1=CC=C(C)C=C1
This item is not returnable. View return policy
18055-70-4
C18H26OSi2
314.58
MFCD09266163
MEQYEVSZYLVDNX-UHFFFAOYSA-N
sym-tetramethyl-di-p-tolyl disiloxane, 1,3-di p-tolyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis p-tolyl-1,1,3,3-tetramethyl disiloxane, disiloxane, 1,1,3,3-tetramethyl-1,3-bis 4-methylphenyl, dimethyl 4-methylphenyl silyl oxy dimethyl 4-methylphenyl silane, amtsi070, 1,3-di-p-tolyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis 4-methylphenyl-1,1,3,3-tetramethyldisiloxane, 1,3-di 4-methylphenyl-1,1,3,3-tetramethyldisiloxane
519510
{[dimethyl(4-methylphenyl)silyl]oxy}dimethyl(4-methylphenyl)silane
CC1=CC=C(C=C1)[Si](C)(C)O[Si](C)(C)C1=CC=C(C)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 314.57
Percent Purity 94%
Chemical Name or Material 1,3-Di(p-tolyl)-1,1,3,3-tetramethyldisiloxane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'