missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1,3-Di-n-butylxanthine, 98%
Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Quantity | 25 g |
Formula Weight | 264.33 |
Percent Purity | 98% |
Chemical Name or Material | 1,3-Di-n-butylxanthine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'