missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclopentanediol, 95%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog Number p-3757358
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
59719-74-3
C5H10O2
102.13
MFCD00001367
NUUPJBRGQCEZSI-UHFFFAOYSA-N
1,3-cyclopentanediol, 1,3-cyclopentanediol, cis, 1,3-cyclopentanediol, mixture of cis and trans, 1,3-cyclopentandiol, 1,4-cyclopentanediol, cyclopentan-1,3-diol, 3-hydroxy cylcopentanol, cyclopentane 1,3-diol, acmc-20ap4n, 1,3-dihydroxycyclopentane
100165
cyclopentane-1,3-diol
C1CC(CC1O)O
This item is not returnable. View return policy
59719-74-3
C5H10O2
102.13
MFCD00001367
NUUPJBRGQCEZSI-UHFFFAOYSA-N
1,3-cyclopentanediol, 1,3-cyclopentanediol, cis, 1,3-cyclopentanediol, mixture of cis and trans, 1,3-cyclopentandiol, 1,4-cyclopentanediol, cyclopentan-1,3-diol, 3-hydroxy cylcopentanol, cyclopentane 1,3-diol, acmc-20ap4n, 1,3-dihydroxycyclopentane
100165
cyclopentane-1,3-diol
C1CC(CC1O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 40.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 80.0°C to 85.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C5H8(OH)2
Refractive Index 1.483
Quantity 1 g
Beilstein 06, III, 4057
Specific Gravity 1.09
Solubility Information Solubility in water: partially soluble.
Formula Weight 102.13
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 1,3-Cyclopentanediol, Mixture of cis and trans
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'