missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Chlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61752.03

Code : NEW

Additional Details : CAS Number : 23751-05-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466756

  • £63.81 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Chlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
314.69
3066591
23751-05-5
MFCD00504706

Specifications

1-(3-Chlorophenyl)-3-[4-(trifluoromethyl)phenyl]urea
23751-05-5
97%
MFCD00504706
1g
Solid
314.69
3066591
Videos
SDS