missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64803.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499156

  • £59.78 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
344.72
MFCD06105843

Specifications

1-(3-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
97%
344.72
Solid
1g
MFCD06105843
Videos
SDS