Alfa Aesar™ 1-(3-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64803.MD

UNSPSC: 12352116

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15419166

Quantity Price
1 £ 14.95 / 250mg
Estimated Shipment date
from Supplier 12-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-(3-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Formula Weight 344.72
MDL Number MFCD06105843

Specification

Formula Weight 344.72
Quantity 250mg
Assay 97%
Chemical Name or Material 1-(3-Chloro-4-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
MDL Number MFCD06105843
Physical Form Solid
Documentation

Technical documents