Alfa Aesar™ 1-(3-Chloro-4-methylphenyl)-3-(4-fluorophenyl)urea, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H62687.06

5GR 1-(3-Chloro-4-methylphenyl)-3-(4-fluorophenyl)urea, 97% 5g

UNSPSC: 12352116

Code: NEW

Additional Details:
Additional Details: Weight: 0.00500kg


Quantity 5g

Product Code. 15427736

Quantity Price
1 £ 251.22 / 5g
Estimated Shipment date
from Supplier 30-01-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-(3-Chloro-4-methylphenyl)-3-(4-fluorophenyl)urea
Formula Weight 278.71
MDL Number MFCD03760989

Specification

Formula Weight 278.71
Chemical Name or Material 1-(3-Chloro-4-methylphenyl)-3-(4-fluorophenyl)urea
Physical Form Solid
Assay 97%
Quantity 5g
MDL Number MFCD03760989