missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chloro-4-fluorophenyl)piperidine, 98%, Thermo Scientific™

Catalog Number 15497878
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1033201-89-6
C11H13ClFN
213.68
MFCD10699654
FOWVTJMHLJVBHP-UHFFFAOYSA-N
1-3-chloro-4-fluorophenyl piperidine
46738781
1-(3-chloro-4-fluorophenyl)piperidine
FC1=C(Cl)C=C(C=C1)N1CCCCC1
This item is not returnable. View return policy
1033201-89-6
C11H13ClFN
213.68
MFCD10699654
FOWVTJMHLJVBHP-UHFFFAOYSA-N
1-3-chloro-4-fluorophenyl piperidine
46738781
1-(3-chloro-4-fluorophenyl)piperidine
FC1=C(Cl)C=C(C=C1)N1CCCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Formula Weight 213.68
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-(3-Chloro-4-fluorophenyl)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'