missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Chloro-4-fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64223.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15438786

  • £54.38 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Chloro-4-fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
348.68
MFCD05990710

Specifications

1-(3-Chloro-4-fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
348.68
1g
97%
MFCD05990710
Videos
SDS