missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Chloro-4-fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64223.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15448786

  • £271.36 / 5g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Chloro-4-fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
348.68
MFCD05990710

Specifications

1-(3-Chloro-4-fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
348.68
5g
97%
MFCD05990710
Videos
SDS