missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Chloro-2-methylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H65001.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15499306

  • £85.44 / 1g


Estimated Shipment date
from Supplier 21-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Chloro-2-methylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
363.16
MFCD04287936
UN3077

Specifications

1-(3-Chloro-2-methylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
97%
1g
MFCD04287936
Solid
Off-white to pale gray
363.16
UN3077
Videos
SDS