missing translation for 'onlineSavingsMsg'
Learn More

(±)-1,3-Butanediol, 99%, Extra Pure, Thermo Scientific Chemicals

Catalog Number 10559990
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
107-88-0
C4H10O2
90.12
MFCD00004554
PUPZLCDOIYMWBV-UHFFFAOYSA-N
1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol
7896
CHEBI:52683
butane-1,3-diol
CC(CCO)O
This item is not returnable. View return policy
107-88-0
C4H10O2
90.12
MFCD00004554
PUPZLCDOIYMWBV-UHFFFAOYSA-N
1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol
7896
CHEBI:52683
butane-1,3-diol
CC(CCO)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -54.0°C
Color Brown to Colorless
Density 1.0100g/mL
Boiling Point 203.0°C to 204.0°C
Flash Point 109°C
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2CH2OH
Refractive Index 1.4385 to 1.4405
Quantity 2.5 L
Beilstein 01, 477
Merck Index 15, 1570
Specific Gravity 1.01
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, methyl ethyl ketone, ethanol, slightly soluble in ether, practically insoluble in aliphatic hydrocarbons,, benzene, toluene and ccl4.
Viscosity 24.6 mm2/s (50°C)
Formula Weight 90.12
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material (±)-1,3-Butanediol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'