missing translation for 'onlineSavingsMsg'
Learn More

1-(3-bromopropoxy)-2-nitrobenzene, 90+%, Thermo Scientific™

Catalog Number 10649964
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
104147-69-5
MFCD00596660
HPZBIRIHQKSGGT-UHFFFAOYSA-N
1-3-bromopropoxy-2-nitrobenzene, 1-3-bromo-propoxy-2-nitro-benzene
7172300
1-(3-bromopropoxy)-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])OCCCBr
This item is not returnable. View return policy
104147-69-5
MFCD00596660
HPZBIRIHQKSGGT-UHFFFAOYSA-N
1-3-bromopropoxy-2-nitrobenzene, 1-3-bromo-propoxy-2-nitro-benzene
7172300
1-(3-bromopropoxy)-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])OCCCBr
CAS Min % 90.0
CAS Max % 100.0
Melting Point 36°C to 39°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 90+%
Packaging Glass bottle
Molecular Formula C9H10BrNO3
Linear Formula NO2C6H4O(CH2)3Br
Quantity 25 g
Molecular Weight (g/mol) 260.087
Formula Weight 260.09
Percent Purity ≥90.0%
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Bromopropoxy)-2-nitrobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-(3-Bromopropoxy)-2-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'