missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(3-Bromophenylsulfonyl)-3-methylpiperidine, 97%

Brand:  Alfa Aesar™ H60793.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477288

  • £358.49 / 1g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(3-Bromophenylsulfonyl)-3-methylpiperidine
C12H16BrNO2S
318.23
MFCD12515196

Specifications

1-(3-Bromophenylsulfonyl)-3-methylpiperidine
C12H16BrNO2S
MFCD12515196
97%
318.23
1g
Videos
SDS