missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Bis(trimethylsilyl)benzene

Brand:  Alfa Aesar™ 017542.06

Code : NEW

Additional Details : CAS Number : 2060-89-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11323377

  • £261.82 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

1,3-Bis(trimethylsilyl)benzene
2060-89-1
222.48
MFCD00015591

Specifications

1,3-Bis(trimethylsilyl)benzene
Liquid
222.48
120°C (25mmHg)
>60° (140°F)
2060-89-1
1.482
MFCD00015591
5g
Videos
SDS